Aluminium oxide with gypsum for TLC

Synonyms: Aluminium(III) oxideAlumina C

46113-22 46113-36
AA46113-36EA 87.71 USD
AA46113-36 AA46113-22
Aluminium oxide with gypsum for TLC
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR