Aluminium oxide for decolorizing

Synonyms: Aluminium(III) oxideAlumina C

46066-30 46066-A1
AA46066-A1EA 134.25 USD
AA46066-A1 AA46066-30
Aluminium oxide for decolorizing
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR