Aluminium oxide basic for TLC
Supplier: Thermo Scientific Chemicals
046001-36
046001-22
AA46001-36EA
147.99
USD
AA46001-36
AA46001-22
Aluminium oxide basic for TLC
Aluminium oxide basic
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2980 °C Melting Pt: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...