Palladium 5% on activated carbon paste (50% water) 5R394
045921-06
045921-14
AA45921-14EA
401.99
USD
AA45921-14
AA45921-06
Palladium 5% on activated carbon paste (50% water) 5R394
Palladium
Formula:
Pd MW: 106.420 g/mol Density: 1.5 Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...