Zinc selenide, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 2 mm (0.08 in)
|
Danger
|
Synonyms:
Zinc (II) selenide
45885-KS
AA45885-KSEA
480.86
USD
AA45885-KS
Zinc selenide, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 2 mm (0.08 in)
Zinc selenide
Formula:
SeZn MW: 144.35 g/mol Melting Pt: 1793 °C Density: 5.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS Number: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 12,10290 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...