Zinc selenide, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 1 mm (0.04 in)

Danger

Synonyms: Zinc (II) selenide

45834-KS
AA45834-KSEA 616.86 USD
AA45834-KS
Zinc selenide, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 1 mm (0.04 in)
Zinc selenide
Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793 °C
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,10290

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR