Silica gel, Pore size: 80 Å ≥99.99%, spherical 20 µm APS for preparative chromatography
|
Warning
|
45830-14
45830-22
45830-06
AA45830-22EA
289.71
USD
AA45830-22
AA45830-14
AA45830-06
Silica gel, Pore size: 80 Å ≥99.99%, spherical 20 µm APS for preparative chromatography
Silica gel
Formula:
SiO₂ MW: 60.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...