Barium metaborate monohydrate

Warning

45823-A7 45823-A1
AA45823-A1EA 43.47 USD
AA45823-A1 AA45823-A7
Barium metaborate monohydrate
Barium metaborate monohydrate
Formula: B₂BaO₄·H₂O
MW: 240.97 g/mol (222.96 anhy)
Melting Pt: >900 °C
Density: 3,35
Storage Temperature: Ambient
MDL Number: MFCD00046182
CAS Number: 19004-06-9
EINECS: 237-222-4
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR