Tungsten(V) ethoxide 1,2-dimethoxyethane adduct ≥99%

Danger

45659-14 45659-06
AA45659-06EA 54.6 USD
AA45659-06 AA45659-14
Tungsten(V) ethoxide 1,2-dimethoxyethane adduct ≥99%
Tungsten(V) ethoxide 1,2-dimethoxyethane adduct
Formula: C₁₀H₂₅O₅W·C₄H₁₀O₂
Boiling Pt: Dec.
Melting Pt: <0 °C
Storage Temperature: Ambient
MDL Number: MFCD16294494
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR