Sodium isopropoxide isopropanol adduct ≥95%

Danger

45648-06 45648-22
AA45648-06EA 35.27 USD
AA45648-06 AA45648-22
Sodium isopropoxide isopropanol adduct ≥95%
Sodium isopropoxide isopropanol adduct
Formula: NaOC₃H₇·C₃H₈O
Melting Pt: 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD12547728
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR