Gallium arsenide, Optical Window, Diameter 50.8 mm (2.0 in), Thickness 2 mm (0.08 in)
|
Danger
|
45636-KS
AA45636-KSEA
1351.98
USD
AA45636-KS
Gallium arsenide, Optical Window, Diameter 50.8 mm (2.0 in), Thickness 2 mm (0.08 in)
Gallium arsenide
Formula:
AsGa MW: 144.64 g/mol Melting Pt: 1238 °C Density: 5.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011017 CAS Number: 1303-00-0 EINECS: 215-114-8 UN: 1557 ADR: 6.1,II Merck Index: 12,04365 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...