Silicon nitride amorphous ≥98.5%, nanopowder
Supplier: Thermo Scientific Chemicals
Synonyms:
Silicon(IV) nitride
045611-06
045611-22
045611-14
AA45611-06EA
50.59
USD
AA45611-06
AA45611-22
AA45611-14
Silicon nitride amorphous ≥98.5%, nanopowder
Silicon nitride
Formula:
N₄Si₃ MW: 140.28 g/mol Melting Pt: 1900 °C (subl.) Density: 3.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011230 CAS Number: 12033-89-5 EINECS: 234-796-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...