Palladium hydroxide (5% Pd) on carbon (Pearlman's catalyst) wetted with ca. 50% water, powder Type A403002-5
Supplier: Thermo Scientific Chemicals
45576-04
45576-09
AA45576-04EA
175.99
USD
AA45576-04
AA45576-09
Palladium hydroxide (5% Pd) on carbon (Pearlman's catalyst) wetted with ca. 50% water, powder Type A403002-5
Palladium hydroxide
Formula:
H₂O₂Pd Storage Temperature: Ambient |
CAS Number:
12135-22-7 EINECS: 235-219-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...