Barium dodecairon nonadecaoxide

Supplier: Thermo Scientific Chemicals
Warning

045552-14 045552-22 045552-06
AA45552-06EA 26.89 USD
AA45552-06 AA45552-22 AA45552-14
Barium dodecairon nonadecaoxide
Barium dodecairon nonadecaoxide
Formula: BaFe₁₂O₁₉
MW: 1111.48 g/mol
Melting Pt: >1315 °C
Storage Temperature: Ambient
MDL Number: MFCD00075637
CAS Number: 12047-11-9
EINECS: 234-974-5
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Form Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR