Boron 10, 10B, plasma standard solution, Specpure®, 10B 100µg/ml, Thermo Fisher Scientific

45530-AD
AA45530-ADEA 161.8 USD
AA45530-AD
Boron 10, 10B, plasma standard solution, Specpure®, 10B 100µg/ml, Thermo Fisher Scientific
Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
Density: 1.0
Boiling Point: 100°
UN No.: 3264
MDL No.: MFCD00143869
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR