Nickel sulfide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel disulphane
, Nickel disulfane
, Nickel subsulphide
, Nickel subsulfide
045498-06
45498-14
AA45498-06EA
197.99
USD
AA45498-06
AA45498-14
Nickel sulfide ≥99% (metals basis)
Nickel sulfide
Formula:
Ni₃S₂ MW: 240.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00217044 CAS Number: 12035-72-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...