Tin 122, 122Sn, Plasma Standard Solution, Specpure®, 122 Sn 10 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
45287-AD
AA45287-ADEA 138.93 USD
AA45287-AD
Tin 122, 122Sn, Plasma Standard Solution, Specpure®, 122 Sn 10 µg/ml, Thermo Scientific
Molecular Formula: Matrix: 2% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00084602
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR