Chromium 50, 50Cr, plasma standard solution, Specpure®, 50Cr 10µg/ml

Supplier: Alfa Aesar
Specpure®
45275-AD
AA45275-ADEA 211.22 USD
AA45275-AD
Chromium 50, 50Cr, plasma standard solution, Specpure®, 50Cr 10µg/ml
Standards Cationic Standards Chromium Standards
Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00190543
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR