Nafion®, membrane, Nafion® NRE-212, Thickness 0.05 mm ≥0.92 meq/g exchange capacity

45036-4X 45036-RF 45036-VA
AA45036-4XEA 856.1 USD
AA45036-4X AA45036-RF AA45036-VA
Nafion®, membrane, Nafion® NRE-212, Thickness 0.05 mm ≥0.92 meq/g exchange capacity
Nafion®

Nafion® NRE-212 perfluorosulfonic acid-PTFE copolymer, nominally 50 microns thick, standard exchange capacity 0.92meq/g. Membrane is positioned between a backing film and a cover sheet.

Formula: C₉HF₁₇O₅S
MW: 544.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00132790
CAS Number: 31175-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR