Stainless steel (Fe:Cr:Ni:Mo; 68:17:12:3 wt%), gauze woven from wire 200 mesh woven from 0.05mm (0.002in) Type 316
Supplier: Thermo Scientific Chemicals
45002-RU
45002-RR
45002-CH
AA45002-CHEA
136.99
USD
AA45002-CH
AA45002-RR
AA45002-RU
Stainless steel (Fe:Cr:Ni:Mo; 68:17:12:3 wt%), gauze woven from wire 200 mesh woven from 0.05mm (0.002in) Type 316
Stainless steel
Open area: 36%; Width of opening: 0.005in