Tantalum disilicide

Supplier: Thermo Scientific Chemicals

Synonyms: Tantalum silicide

44965-09 44965-18
AA44965-18EA 265.99 USD
AA44965-18 AA44965-09
Tantalum disilicide
Tantalum disilicide
Formula: Si₂Ta
MW: 237.118 g/mol
Melting Pt: 2200 °C
Storage Temperature: Ambient
MDL Number: MFCD00049564
CAS Number: 12039-79-1
EINECS: 234-902-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR