Sodium-silica gel 35-40% alkali metal in silica gel Stage II

Danger

44894-14 44894-06
AA44894-06EA 55.02 USD
AA44894-06 AA44894-14
Sodium-silica gel 35-40% alkali metal in silica gel Stage II
Sodium-silica gel

Applications: Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds. Also ideal for reduction of water to produce hydrogen gas, solvent drying and anti-oxidant

Density: 0,7
Storage Temperature: Ambient
MDL Number: MFCD01941718
UN: 2813
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR