Allyl acrylate ≥95% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acrylic Acid Allyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

44463-14
AA44463-14EA 373.99 USD
AA44463-14
Allyl acrylate ≥95% stabilized
Allyl acrylate
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 46…47 °C (40 mmHg)
Density: 0.935
Flash Pt: 32 °C (89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014949
CAS Number: 999-55-3
EINECS: 213-662-2
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR