Platinum : ruthenium Pt:Ru (40:20)% on carbon black, HiSPEC® 10000

Danger

44172-04 44172-09 44172-09 44172-04
AA44172-04EA 385.67 USD
CAAA44172-04 AA44172-04 AA44172-09 CAAA44172-09
Platinum : ruthenium Pt:Ru (40:20)% on carbon black, HiSPEC® 10000
Platinum : ruthenium
Formula: Pt:Ru
Storage Temperature: Ambient
MDL Number: MFCD00798552
CAS Number: 7440-06-4
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR