2,2-Diphenyl-1-picrylhydrazyl ≥95% free radical

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2-Diphenyl-1-(2,4,6-trinitrophenyl)hydrazylDPPH1,1-Diphenyl-2-picrylhydrazyl

44150-03 44150-MD 44150-03 44150-MD
AA44150-03EA 216.93 USD
AA44150-03 CAAA44150-MD CAAA44150-03 AA44150-MD
2,2-Diphenyl-1-picrylhydrazyl ≥95% free radical
2,2-Diphenyl-1-picrylhydrazyl

2,2-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger. It serves as a general antioxidant detector. It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals. Further, it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol.


Appearance: Green to black powder

Formula: C₁₈H₁₂N₅O₆
MW: 394.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007231
CAS Number: 1898-66-4
EINECS: 217-591-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR