Nickel plating solution, electroless ammonia-free
|
Danger
|
44070-AK
44070-AK
44070-K2
44070-K2
AA44070-AKEA
33.69
USD
CAAA44070-AK
AA44070-AK
CAAA44070-K2
AA44070-K2
Nickel plating solution, electroless ammonia-free
Nickel plating solution
Applications: For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
Storage Temperature:
Ambient |
MDL Number:
MFCD00011137 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...