Tetraammineplatinum(II) hydrogen phosphate solution (2% Pt) (w/w)

Danger

44047-NH 44047-NB
AA44047-NHEA 218.67 USD
AA44047-NH AA44047-NB
Tetraammineplatinum(II) hydrogen phosphate solution (2% Pt) (w/w)
Tetraammineplatinum(II) hydrogen phosphate
Formula: H₁₃N₄O₄PPt
MW: 359.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788258
CAS Number: 127733-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR