Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
|
Danger
|
Synonyms:
Bis(norbornadiene)rhodium(I) tetrafluoroborate
, Bis(norbornadiene)rhodium (I) tetrafluoroborate
44036-06
44036-03
44036-03
44036-06
AA44036-06EA
2976.99
USD
AA44036-06
CAAA44036-06
AA44036-03
CAAA44036-03
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Formula:
C₁₄H₁₆BF₄Rh MW: 365,92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00671775 CAS Number: 36620-11-8 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...