Barium-2-ethylhexanoate 20% (w/w) in 2-ethylhexanoic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
44021-30
44021-30
44021-18
44021-18
44021-09
44021-09
AA44021-09EA
48
USD
AA44021-09
AA44021-18
CAAA44021-09
CAAA44021-18
AA44021-30
CAAA44021-30
Barium-2-ethylhexanoate 20% (w/w) in 2-ethylhexanoic acid
Barium 2-ethylhexanoate
Formula:
C₁₆H₃₀BaO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00058696 CAS Number: 2457-01-4 EINECS: 219-535-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...