C-276 alloy, foil, Thickness 0.2 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43994-GH
43994-GH
AA43994-GHEA
133
USD
CAAA43994-GH
AA43994-GH
C-276 alloy, foil, Thickness 0.2 mm (0.008 in)
C-276 alloy
High strength, corrosion resistant alloy
Ni:Mo:Cr:Fe:W;Co:Mn; 57.5:15.5:15.5:6:3.5:1.5:0.5 approximate wt%