5-Ethylidene-2-norbornene (endo + exo mixture) ≥98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Ethylidenebicyclo[2.2.1]hept-2-ene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 100 ppm
043972-AE
43972-AC
43972-AC
43972-AE
43972-AP
43972-AP
AA43972-APEA
121.99
USD
AA43972-AP
CAAA43972-AP
AA43972-AC
CAAA43972-AE
CAAA43972-AC
AA43972-AE
5-Ethylidene-2-norbornene (endo + exo mixture) ≥98% stabilized
5-Ethylidene-2-norbornene
Formula:
C₉H₁₂ MW: 120.20 g/mol Boiling Pt: 146 °C Density: 0.89 Flash Pt: 38 °C (101 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00167576 CAS Number: 16219-75-3 EINECS: 240-347-7 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...