Dibenzofuran-2-sulfonic acid hydrate ≥97%

Warning

43899-03 43899-03
AA43899-03EA 69.7 USD
CAAA43899-03 AA43899-03
Dibenzofuran-2-sulfonic acid hydrate ≥97%
Dibenzofuran-2-sulfonic acid hydrate
Formula: C₁₂H₈O₄S.xH₂O
MW: 248.24 g/mol (anhy)
Melting Pt: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00043577
CAS Number: 83863-63-2
EINECS: 281-128-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR