Polyimide, resin

Warning

43656-18 43656-18 43656-30 43656-09 43656-09 43656-30
AA43656-09EA 47.1 USD
CAAA43656-18 AA43656-09 AA43656-18 CAAA43656-09 AA43656-30 CAAA43656-30
Polyimide, resin
Polyimide

Soluble thermoplastic polyimide for high temperature adhesives and composites
Tg÷305°

Formula: C₃₅H₂₈N₂O₇
Melting Pt: >300 °C
Density: 1,2
Flash Pt: >93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD01868102
CAS Number: 62929-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR