Rhodium 1% on polyethylenimine/SiO2, powder

Warning

43409-06 43409-14 43409-14 43409-06
AA43409-14EA 642.99 USD
CAAA43409-06 AA43409-14 CAAA43409-14 AA43409-06
Rhodium 1% on polyethylenimine/SiO2, powder
Rhodium
Formula: Rh
MW: 102.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR