Chloro(decyl)dimethylsilane ≥97%

Danger

Synonyms: 1-(Chlorodimethylsilyl)decaneDecyldimethylchlorosilane

42769-18 42769-18
AA42769-18EA 243.68 USD
AA42769-18 CAAA42769-18
Chloro(decyl)dimethylsilane ≥97%
Chloro(decyl)dimethylsilane
Formula: C₁₂H₂₇ClSi
MW: 234.88 g/mol
Boiling Pt: 98 °C (2 mmHg)
Density: 0,866
Flash Pt: 137 °C (279 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054168
CAS Number: 38051-57-9
EINECS: 253-761-8
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR