Cholesterol-2,4-dichlorobenzoate ≥96%

Synonyms: Cholesteryl 2,4-Dichlorobenzoate

42700-09 42700-30 42700-30 42700-09 42700-18 42700-18
AA42700-30EA 176.49 USD
CAAA42700-30 AA42700-30 CAAA42700-09 CAAA42700-18 AA42700-18 AA42700-09
Cholesterol-2,4-dichlorobenzoate ≥96%
Cholesterol-2,4-dichlorobenzoate

Transition temperatures: Cholesteric-isotropic: >210°C; residual cholesterol: <0.7%

Soluble in THF

Formula: C₃₄H₄₈Cl₂O₂
MW: 559.66 g/mol
Melting Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD07776849
CAS Number: 32832-01-2
EINECS: 251-248-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR