Potassium methanolate 5% (w/v) in methanol

Danger

42551-AC 42551-AC
AA42551-ACEA 53.13 USD
AA42551-AC CAAA42551-AC
Potassium methanolate 5% (w/v) in methanol
Potassium methanolate
Formula: CH₃KO
MW: 70.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012416
CAS Number: 865-33-8
UN: 3274
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR