Ruby Spheres, Grade 25, Thermo Scientific

Supplier: Thermo Scientific
42044-KN 42078-KN 42042-KJ 42045-KH 42077-KN 42084-KH 42081-KJ 42081-KM 42041-KM 42082-KJ 42073-KJ 42042-KN 42078-KM 42073-KM 42073-KN 42081-KN 42083-KH 42045-KK 42077-KJ 42072-KN 42044-KJ 42079-KN 42043-KM 42044-KM 42041-KJ 42045-KD 42041-KN 42083-KK 42079-KJ 42075-KJ 42078-KJ 42082-KN 42084-KK 42083-KD 42084-KD 42043-KJ 42042-KM 42043-KN 42075-KN 42075-KM 42079-KM 42077-KM 42082-KM
AA42045-KKEA 282.57 USD
AA42045-KK AA42072-KN AA42043-KN AA42043-KM AA42083-KD AA42041-KJ AA42045-KD AA42082-KN AA42043-KJ AA42082-KM AA42083-KH AA42045-KH AA42082-KJ AA42042-KN AA42079-KN AA42077-KJ AA42073-KJ AA42079-KJ AA42075-KM AA42077-KN AA42077-KM AA42075-KJ AA42079-KM AA42073-KN AA42044-KJ AA42073-KM AA42075-KN AA42044-KM AA42044-KN AA42084-KD AA42042-KM AA42081-KN AA42042-KJ AA42081-KM AA42084-KH AA42081-KJ AA42084-KK AA42083-KK AA42041-KM AA42041-KN AA42078-KM AA42078-KN AA42078-KJ
Ruby Spheres, Grade 25, Thermo Scientific
Specimens Mineral Specimens Gems and Tumbled Stones
  • Diameter tolerance per half: ±0.000025"
  • V block out of round in 120° angle: ±0.000025"
  • Basic diameter tolerance: ±0.0001"
  • Maximum surface roughness (arithmetic average): 1.5 micro-inch

MDL No.: MFCD00003424
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR