Samarium(III) phosphate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

041992-09 41992-09
AA41992-09EA 128.99 USD
AA41992-09 CAAA41992-09
Samarium(III) phosphate hydrate ≥99.9% (REO, rare earth oxide basis)
Samarium(III) phosphate hydrate
Formula: SmPO₄.xH₂O
MW: 246.41 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD05665148
CAS Number: 14913-18-9
EINECS: 236-698-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR