Polystyrene standard, M.W. 30,000, Mw/Mn 1.06, Alfa Aesar

41930-06 41930-03 41930-06
AA41930-06EA 227.08 USD
CAAA41930-06 AA41930-06 CAAA41930-03
Polystyrene standard, M.W. 30,000, Mw/Mn 1.06, Alfa Aesar
CAS No.: 9003-53-6
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00084450
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR