Butylmagnesium chloride 1.5-2.5 M in THF

Danger

Synonyms: n-Butylmagnesium chloride

Packaged under Argon in resealable ChemSeal™ bottles.

41676-AD 41676-AK
AA41676-ADEA 61.86 USD
AA41676-AD AA41676-AK
Butylmagnesium chloride 1.5-2.5 M in THF
Butylmagnesium chloride

Applications: Grignard reagent


Applications: Grignard reagent

Formula: C₄H₉ClMg
MW: 116.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000475
CAS Number: 693-04-9
EINECS: 211-739-5
UN: 3399
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR