Crucibles, Conical, Flat Base, Zirconium Oxide, Thermo Scientific
Supplier: Thermo Scientific
41291-KF
AA41291-KFEA 663.78 USD
AA41291-KF
Crucibles, Conical, Flat Base, Zirconium Oxide, Thermo Scientific
These crucibles are manufactured from yttria stabilized zirconium oxide
  • Increased working temperature characteristics
  • Offer useful alternatives to aluminium
They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.