Methylmercury hydroxide 1 M in water

Danger

Synonyms: Methylmercury(II) hydroxide

Packaged under argon in resealable ChemSeal™ bottles.

41253-AB 41253-AB 41253-XF 41253-AD 41253-XF
AA41253-XFEA 52.26 USD
CAAA41253-AB AA41253-XF AA41253-AB CAAA41253-AD CAAA41253-XF
Methylmercury hydroxide 1 M in water
Methylmercury hydroxide
Formula: CH₄HgO
MW: 232.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013924
CAS Number: 1184-57-2
EINECS: 214-667-2
UN: 2024
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR