Molybdenum disilicide ≥99.5% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Synonyms: Molybdenum silicide

41076-KS
CAAA41076-KS
Molybdenum disilicide ≥99.5% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Molybdenum disilicide

Applications: Magnetron sputtering source

Formula: MoSi₂
MW: 152.11 g/mol
Melting Pt: 1870…2030 °C
Density: 6,31
Storage Temperature: Ambient
MDL Number: MFCD00151535
CAS Number: 12136-78-6
EINECS: 234-786-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR