Lead zirconium titanium oxide ≥99.9% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)

Danger

Synonyms: Lead zirconate titanate

41070-KS
CAAA41070-KS
Lead zirconium titanium oxide ≥99.9% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)
Lead zirconium titanium oxide
Formula: PbZr(1-x)TixO₃
Storage Temperature: Ambient
MDL Number: MFCD00212270
CAS Number: 12626-81-2
EINECS: 235-727-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR