Lead zirconium titanium oxide ≥99.9% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Danger

Synonyms: Lead zirconate titanate

41068-KS 41068-KS
AA41068-KSEA 734.82 USD
AA41068-KS CAAA41068-KS
Lead zirconium titanium oxide ≥99.9% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Lead zirconium titanium oxide

Applications: Magnetron sputtering source

Formula: PbZr(1-x)TixO₃
Storage Temperature: Ambient
MDL Number: MFCD00212270
CAS Number: 12626-81-2
EINECS: 235-727-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR