Chromium disilicide ≥99.5% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 6.35 mm (0.250 in)

Danger

Synonyms: Chromium silicide

41061-KS
CAAA41061-KS
Chromium disilicide ≥99.5% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 6.35 mm (0.250 in)
Chromium disilicide
Formula: CrSi₂
Storage Temperature: Ambient
MDL Number: MFCD00168070
CAS Number: 12018-09-6
EINECS: 234-633-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR