Chromium disilicide ≥99.5% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Danger

Synonyms: Chromium silicide

41060-KS
CAAA41060-KS
Chromium disilicide ≥99.5% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Chromium disilicide

Applications: Magnetron sputtering source

Formula: CrSi₂
Storage Temperature: Ambient
MDL Number: MFCD00168070
CAS Number: 12018-09-6
EINECS: 234-633-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR