Nafion® NR50, pellets super acidic ion exchange resin catalyst

Warning

40802-09 40802-18 40802-09 40802-18
AA40802-09EA 178.97 USD
AA40802-09 CAAA40802-18 AA40802-18 CAAA40802-09
Nafion® NR50, pellets super acidic ion exchange resin catalyst
Nafion® NR50

Sensitivity: Hygroscopic
Appearance: White to pale brown
Applications: Super acid catalyst

Formula: C₉HF₁₇O₅S
Melting Pt: 200 °C
Density: 2.1
Storage Temperature: Ambient
MDL Number: MFCD00132790
CAS Number: 31175-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR