Scandium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

40579-NA 040579-NA
AA40579-NAEA 88.29 USD
CAAA40579-NA AA40579-NA
Scandium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Scandium (III) perchlorate
Formula: Cl₃O₁₂Sc
MW: 451.42 g/mol (343.30 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00243289
CAS Number: 14066-05-8
EINECS: 237-914-6
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR